Learn More
ACROBiosystems Recombinant Protein;Human CD96 / TACTILE Protein, His Tag;HEK293;100UG;TAE,CD96,Tactile
Catalog No. 501658239
Encompass_Preferred
Recombinant Protein;Human CD96 / TACTILE Protein, His Tag;100UG;Human CD96, His Tag (TAE-H52H0) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 503 (Accession # P40200-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.4 kDa. The protein migrates as 95-130 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD155, Fc Tag (Cat. No. CD5-H5251) at 5 μg/mL (100 μL/well) can bind Human CD96, His Tag (Cat. No. TAE-H52H0) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-165-8239 Supplier ACROBiosystems Supplier No. TAEH52H0100UG
Recombinant Protein;Human CD96 / TACTILE Protein, His Tag;100UG;Human CD96, His Tag (TAE-H52H0) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 503 (Accession # P40200-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.4 kDa. The protein migrates as 95-130 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD155, Fc Tag (Cat. No. CD5-H5251) at 5 μg/mL (100 μL/well) can bind Human CD96, His Tag (Cat. No. TAE-H52H0) with a linear range of 0.156-1.25 μg/mL (QC tested).