Learn More
ACROBiosystems Recombinant Protein;Human CD96 / TACTILE Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;TAE,CD96,Tactile
Supplier:  ACROBiosystems TAEH52521MG
Encompass_Preferred
Recombinant Protein;Human CD96 / TACTILE Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human CD96, Mouse IgG2a Fc Tag, low endotoxin (TAE-H5252) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 503 (Accession # P40200-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 80.4 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD96, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. TAE-H5252) at 2 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-165-7642