Learn More
Genscript Corporation CD98 His Cynomolgus
Catalog No. 5000059072
Encompass
The type II transmembrane protein CD98 best known as the heavy chain of the heterodimeric amino acid transporters (HAT) is required for the surface expression and basolateral localization of this transporter complex in polarized epithelial cells CD98 also interacts with beta1 integrins resulting in an increase in their affinity for ligand In this study we explored the role of the transmembrane and cytoplasmic domains of CD98 on integrin-dependent cell adhesion and migration in polarized renal epithelial cells
Catalog No. 50-000-59072 Supplier Genscript Corporation Supplier No. Z04129500
The type II transmembrane protein CD98 best known as the heavy chain of the heterodimeric amino acid transporters (HAT) is required for the surface expression and basolateral localization of this transporter complex in polarized epithelial cells CD98 also interacts with beta1 integrins resulting in an increase in their affinity for ligand In this study we explored the role of the transmembrane and cytoplasmic domains of CD98 on integrin-dependent cell adhesion and migration in polarized renal epithelial cells