Learn More
ABclonal Technology CD98/SLC3A2 Rabbit
SDP
Catalog No. 5000008107
Encompass_Preferred
This gene is a member of the solute carrier family and encodes a cell surface transmembrane protein The protein exists as the heavy chain of a heterodimer covalently bound through di-sulfide bonds to one of several possible light chains The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids Alternatively spliced transcript variants encoding different isoforms have been characterized
Catalog No. 50-000-08107 Supplier ABclonal Technology Supplier No. A19880PM20UL
This gene is a member of the solute carrier family and encodes a cell surface transmembrane protein The protein exists as the heavy chain of a heterodimer covalently bound through di-sulfide bonds to one of several possible light chains The encoded transporter plays a role in regulation of intracellular calcium levels and transports L-type amino acids Alternatively spliced transcript variants encoding different isoforms have been characterized