Learn More
ACROBiosystems Recombinant Protein;Human CD98 Protein;HEK293;100UG;CD8,SLC3A2,CD98,4F2,4F2HC,4T2HC,CD98HC,MDU1,NACAE
Supplier:  ACROBiosystems CD8H5247100UG
Encompass_Preferred
Recombinant Protein;Human CD98 Protein;100UG;Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contains AA Arg 206 - Ala 630  (Accession # AAH01061).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 50-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8119