Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4267RBM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: MIC2/1495R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG kappa. Positive Control: MOLT-4 cells. Pancreas or Ewing s sarcoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with or without 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length MIC2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 27-32kDa.Chromosome: Xp22.33. Gene Symbol: CD99. Gene ID: 4267. Ensemble: ENSG00000002586. Uniprot: NP_001308299.
Catalog No. 50-188-4938