Learn More
Bioss CD99 - Extracellular region An
SDP
Supplier:  Bioss BSM70407M
Encompass_Preferred
CD99 (Extracellular region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB IP M026Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65301