Learn More
ACROBiosystems ACROBiosystems Human CD99 / MIC2 Protein, His Tag
Catalog No. 501957487
Encompass_Preferred
Recombinant Protein;1MG;Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # AAH02584).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7487 Supplier ACROBiosystems Supplier No. CD9H52211MG
Recombinant Protein;1MG;Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession # AAH02584).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.9 kDa. The protein migrates as 25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.