Learn More
Enzo Life Sciences CD99 Recombinant monoclonal antibody (MIC2/1495R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3570100
Encompass_Preferred
Clone: MIC2/1495R. Isotype: IgGκ. Immunogen: Recombinant full length human CD99. Species reactivity: Human. Applications: Flow Cytometry, IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2886