Learn More
Genscript Corporation CD99/MIC2 hFc Chimera Human
Catalog No. 5000057662
Encompass
CD99 is a cell surface protein with unique features and only partly defined mechanisms of action This molecule is involved in crucial biological processes including cell adhesion migration death differentiation and diapedesis and it influences processes associated with inflammation immune responses and cancer CD99 is frequently overexpressed in many types of tumors particularly pediatric tumors including Ewing sarcoma and specific subtypes of leukemia
Catalog No. 50-000-57662 Supplier Genscript Corporation Supplier No. Z04242100
CD99 is a cell surface protein with unique features and only partly defined mechanisms of action This molecule is involved in crucial biological processes including cell adhesion migration death differentiation and diapedesis and it influences processes associated with inflammation immune responses and cancer CD99 is frequently overexpressed in many types of tumors particularly pediatric tumors including Ewing sarcoma and specific subtypes of leukemia