Learn More
AdipoGen CDAP
SDP
Catalog No. 502561415
Encompass_Preferred
Chemical. CAS 59016-56-7. Formula C8H10N3 . BF4. MW 235. 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate CDAP is an organic highly effective water soluble cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity. The activation and further carbohydrate modification are produced under mild conditions in an aqueous solvent, an enzyme friendly environment. CDAP can be used with alkaline-sensitive polysaccharides and proteins.
Catalog No. 50-256-1415 Supplier AdipoGen Supplier No. CDXC0235M500
Chemical. CAS 59016-56-7. Formula C8H10N3 . BF4. MW 235. 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate CDAP is an organic highly effective water soluble cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity. The activation and further carbohydrate modification are produced under mild conditions in an aqueous solvent, an enzyme friendly environment. CDAP can be used with alkaline-sensitive polysaccharides and proteins.