Learn More
Enzo Life Sciences CDC (10mg). CAS: 132465-11-3
SDP
Catalog No. 502010610
Encompass_Preferred
Selective 12-lipoxygenase inhibitor. IC50=0.063 µM, 3.33 µM and 1.89 µM for 12-, 15- and 5-LO respectively in isolated enzyme preparations. Purity: ≥98%. Solubility: Soluble in ethanol (5mg/ml) or DMSO (5mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0610 Supplier Enzo Life Sciences Supplier No. BMLEI2110010
Selective 12-lipoxygenase inhibitor. IC50=0.063 µM, 3.33 µM and 1.89 µM for 12-, 15- and 5-LO respectively in isolated enzyme preparations. Purity: ≥98%. Solubility: Soluble in ethanol (5mg/ml) or DMSO (5mg/ml). Long Term Storage: Ambient.