Learn More
Abcam CDC, lipo x ygenase inhibitor, 50MG
Catalog No. 502706523
Encompass_Preferred
MW 321.3 g/mol, Purity >98%. Potent, selective 12-Lipoxygenase (12-LO) inhibitor (IC₅₀ = 63 nM). Also inhibits 15-LO and 5-LO (IC₅₀ values are 3.33 and 1.89 μM respectively). Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6523 Supplier Abcam Supplier No. AB14156050MG
MW 321.3 g/mol, Purity >98%. Potent, selective 12-Lipoxygenase (12-LO) inhibitor (IC₅₀ = 63 nM). Also inhibits 15-LO and 5-LO (IC₅₀ values are 3.33 and 1.89 μM respectively). Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement