Learn More
Aviva Systems Biology Corp CDC2 Antibody - middle region (AVARP01006_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP01006_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDC2. Alternate names: CDC28A, CDK1, DKFZp686L20222, MGC111195, CDC2, P34CDC2ApplicationWB. Species reactivity:
Catalog No. 50-014-54082