Learn More
Aviva Systems Biology Corp CDC2 Antibody - C-terminal region (AVARP03024_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03024_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CDC2. Alternate names: CDC28A, CDK1, DKFZp686L20222, MGC111195, CDC2, P34CDC2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-54085