Learn More
Santa Cruz Biotechnology Cdc2 p34 Antibody (17), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC54
Encompass
Cdc2 p34 Antibody (17) is recommended for detection of Cdc2 p34 of mouse, rat, human and Xenopus origin by WB, IP, IF and IHC(P).
Catalog No. NC1762528