Learn More
Enzo Life Sciences cdc2 substrate (biotinylated) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2720001
Encompass_Preferred
This peptide, (Ac-S)-PGRRRRK-biotin, is a substrate for CDC2 kinase. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1328