Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 991MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CDC20/1102. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Ramos or HeLa cells. Tonsil or gastric carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Cdc20 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 55kDa.Chromosome: 1p34.2. Gene Symbol: CDC20. Gene ID: 991. Ensemble: ENSG00000117399. Uniprot: NP_001246.
Catalog No. 50-188-5393