Learn More
ABclonal Technology CDC20 Rabbit pAb
SDP
Catalog No. 5000105210
Encompass_Preferred
CDC20 appears to act as a regulatory protein interacting with several other proteins at multiple points in the cell cycle. It is required for two microtubule-dependent processes, nuclear movement prior to anaphase and chromosome separation.
Catalog No. 50-001-05210 Supplier ABclonal Technology Supplier No. A156561000UL
CDC20 appears to act as a regulatory protein interacting with several other proteins at multiple points in the cell cycle. It is required for two microtubule-dependent processes, nuclear movement prior to anaphase and chromosome separation.