Learn More
Aviva Systems Biology Corp CDC23 Antibody - C-terminal region (ARP43040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDC23. Alternate names: APC8, CUT23, ANAPC8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58405