Learn More
Aviva Systems Biology Corp CDC25A Antibody - middle region (AVARP03040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC25A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDC25A. Alternate names: CDC25A2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62523