Learn More
Enzo Life Sciences CDC25A monoclonal antibody (DCS-120) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC085E
Encompass_Preferred
Clone: DCS-120. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human cdc25A. Species reactivity: Human, Mouse, Rat. Applications: IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7235