Learn More
Aviva Systems Biology Corp CDC25B Antibody - N-terminal region (AVARP03041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC25B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CDC25B. Alternate names: ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Pig, Rabbit
Catalog No. 50-014-62524