Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdc25b-in-1 | 2374831-10-2 | 98.1% | 321.37 g/mol | C20H19NO3 | 10MG
SDP

Catalog No. 5000239560
Encompass_Preferred

CDC25B-IN-1 is a small-molecule research inhibitor of the cell division cycle 25B (CDC25B) phosphatase, referred to in the literature as compound 4a. It is provided as a 10 mg research sample (CAS 2374831-10-2) for biochemical and cellular studies targeting CDC25B-mediated signaling.

  • Potent CDC25B inhibition with reported Ki of 8.5 μM.
  • High purity suitable for research (≈98.1%).
  • Molecular weight 321.37 g/mol; formula C20H19NO3.
  • Suitable for in vitro biochemical and cell-based assays.
  • Storage under nitrogen recommended; refrigerated at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a small quantity research sample for laboratory use.

Catalog No. 50-002-39560 Supplier Medchemexpress LLC Supplier No. HY12624610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CDC25B-IN-1 is a small-molecule research inhibitor of the cell division cycle 25B (CDC25B) phosphatase, referred to in the literature as compound 4a. It is provided as a 10 mg research sample (CAS 2374831-10-2) for biochemical and cellular studies targeting CDC25B-mediated signaling.

  • Potent CDC25B inhibition with reported Ki of 8.5 μM.
  • High purity suitable for research (≈98.1%).
  • Molecular weight 321.37 g/mol; formula C20H19NO3.
  • Suitable for in vitro biochemical and cell-based assays.
  • Storage under nitrogen recommended; refrigerated at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • Supplied as a small quantity research sample for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.