Learn More
Medchemexpress LLC CDC25B-IN-1 | 2374831-10-2 | 98.1% | 321.37 g/mol | C20H19NO3 | 50MG
SDP
Catalog No. 5000239431
Encompass_Preferred
CDC25B-IN-1 is a small-molecule research inhibitor of the cell division cycle 25B (CDC25B) phosphatase. It has a reported Ki of approximately 8.5 μM, reduces cell proliferation and colony formation, and increases the proportion of cells in the G2/M phase. The compound is supplied at high reported purity and is available in multiple pack sizes, including mg quantities and as a DMSO solution suitable for biochemical and cell-based assays.
  • Potent inhibitor of CDC25B phosphatase with reported Ki ≈ 8.5 μM.
  • Inhibits cell proliferation and colony formation in cellular assays.
  • Causes accumulation of cells in the G2/M phase.
  • Supplied with high reported purity for research use.
  • Available in multiple pack sizes and as a DMSO solution for convenience.
Catalog No. 50-002-39431 Supplier Medchemexpress LLC Supplier No. HY12624650MG
CDC25B-IN-1 is a small-molecule research inhibitor of the cell division cycle 25B (CDC25B) phosphatase. It has a reported Ki of approximately 8.5 μM, reduces cell proliferation and colony formation, and increases the proportion of cells in the G2/M phase. The compound is supplied at high reported purity and is available in multiple pack sizes, including mg quantities and as a DMSO solution suitable for biochemical and cell-based assays.
  • Potent inhibitor of CDC25B phosphatase with reported Ki ≈ 8.5 μM.
  • Inhibits cell proliferation and colony formation in cellular assays.
  • Causes accumulation of cells in the G2/M phase.
  • Supplied with high reported purity for research use.
  • Available in multiple pack sizes and as a DMSO solution for convenience.