Learn More
Medchemexpress LLC HY-126246 5mg Medchemexpress, CDC25B-IN-1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1262465MG
Encompass_Preferred
Medchemexpress, HY-126246 5mg CDC25B-IN-1 CAS: CDC25B-IN-1 (compound 4a) is a potent inhibitor of cell division cycle 25B (CDC25B) phosphatase, with a Ki of 8.5 μM. CDC25B-IN-1 potently inhibits cell proliferation and colony formation, causes an increase of the G2/M phase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9722