Learn More
Aviva Systems Biology Corp CDC25C Antibody - N-terminal region (AVARP03034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC25C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CDC25C. Alternate names: CDC25, PPP1R60ApplicationWB. Species reactivity: Human, Mouse, Rabbit
Catalog No. 50-014-62518