Learn More
Enzo Life Sciences CDC25C (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3660050
Encompass_Preferred
Alternative name: Cell division cycle 25C, Dual specificity phosphatase CDC25C. Formulation: Liquid. In 50mM TRIS/HCl, pH 8.0, 150mM NaCl, 5mM DTT, 0.1mM EDTA, 0.03% Brij 35, 20% glycerol. Source: Produced in E. coli. Full length human CDC25C. Long Term Storage: -80°C.
Catalog No. 50-201-2001