Learn More
Aviva Systems Biology Corp CDC26 Antibody - C-terminal region (ARP59256_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59256_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC26. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CDC26. Alternate names: C9orf17, APC12, ANAPC12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-634