Learn More
Aviva Systems Biology Corp CDC34 Antibody - middle region (AVARP03048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC34. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDC34. Alternate names: E2-CDC34, UBC3, UBE2R1, UBCH3ApplicationWB, IHC. Species reactivity: Cow, Dog, Rat
Catalog No. 50-014-62528