Learn More
Aviva Systems Biology Corp CDC37L1 Antibody - C-terminal region (ARP62437_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC37L1. It was validated on Western Blot. . Alternate names: CDC37B, FLJ20639, HARC, RP11-6J24.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-684