Learn More
Aviva Systems Biology Corp CDC40 Antibody - N-terminal region (ARP64500_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64500_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC40. It was validated on Western Blot. . Alternate names: EHB3, FLJ10564, MGC102802, PRP17, PRPF17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-276