Learn More
Medchemexpress LLC Cdc42-in-1 | 1222513-26-9 | MFCD28009507 | 99.8% | 486.38 g·mol⁻1 | C22H20BrN3O3S | 5 MG
SDP
Catalog No. 5000222111
Encompass_Preferred
CID44216842 (Cdc42-IN-1) is a small-molecule inhibitor selective for the Rho GTPase Cdc42. It blocks guanine nucleotide (GTP/GDP) binding with low-micromolar potency and is provided as a high-purity solid for use as a biochemical and cellular research probe.
  • Inhibits Cdc42 GTP/GDP binding with low-micromolar potency.
  • Supplied as a high-purity solid suitable for biochemical assays.
  • Molecular formula C22H20BrN3O3S and molecular weight 486.38 g·mol⁻1.
  • Available in small research quantities such as 5 MG.
  • Store powder at -20°C for long-term stability; limited stability in solution.
Catalog No. 50-002-22111 Supplier Medchemexpress LLC Supplier No. HY1363795MG
CID44216842 (Cdc42-IN-1) is a small-molecule inhibitor selective for the Rho GTPase Cdc42. It blocks guanine nucleotide (GTP/GDP) binding with low-micromolar potency and is provided as a high-purity solid for use as a biochemical and cellular research probe.
  • Inhibits Cdc42 GTP/GDP binding with low-micromolar potency.
  • Supplied as a high-purity solid suitable for biochemical assays.
  • Molecular formula C22H20BrN3O3S and molecular weight 486.38 g·mol⁻1.
  • Available in small research quantities such as 5 MG.
  • Store powder at -20°C for long-term stability; limited stability in solution.