Learn More
Aviva Systems Biology Corp CDC42BPB Antibody - C-terminal region (ARP65418_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65418_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC42BPB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CDC42BPB. Alternate names: KIAA1124, MRCKBApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-496