Learn More
Aviva Systems Biology Corp Cdc42ep4 Antibody - middle region (ARP54840_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54840_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cdc42ep4. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-970