Learn More
Aviva Systems Biology Corp CDC42EP5 Antibody - middle region (ARP52797_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52797_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC42EP5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDC42EP5. Alternate names: Borg3, CEP5, MGC21945, MGC71153ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-60834