Learn More
Aviva Systems Biology Corp CDC5L Antibody - middle region (ARP40347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC5L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDC5L. Alternate names: CEF1, KIAA0432, PCDC5RP, dJ319D22.1, hCDC5, CDC5, CDC5-LIKEApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57403