Learn More
Hach Company Radiometer Analytical CDC641T Conductivity Cell with Temperature Sensor (2-pole Pt sensor, k = 0.85 cm-1, glass body)
Catalog No. 501997532 Shop All Hach Company Products
Encompass
The Radiometer CDC641T is a 2-pole conductivity cell with platinized, platinum sensor (k = 0.85 cm-1), glass body and integrated temperature sensor. The CDC641T sensor's 2 platinum plate design is ideal for routine conductivity measurements or for continuous measurement in various aqueous and non-aqueous media. Additionally, the CDC641T meets requirements of EP 2.2.38. The cell has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M022 for a version of the CDC641T with RJ45 plug, compatible for use with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7532 Supplier Hach Company Supplier No. B15B001
The Radiometer CDC641T is a 2-pole conductivity cell with platinized, platinum sensor (k = 0.85 cm-1), glass body and integrated temperature sensor. The CDC641T sensor's 2 platinum plate design is ideal for routine conductivity measurements or for continuous measurement in various aqueous and non-aqueous media. Additionally, the CDC641T meets requirements of EP 2.2.38. The cell has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M022 for a version of the CDC641T with RJ45 plug, compatible for use with Hach sensION+ benchtop conductivity meters.