Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDC7/DBF4, GST Tag Recombinant Protein

Catalog No. PV6274
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6274 10 μg
PV6275 100 μg
PV6276 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6274 Supplier Invitrogen™ Supplier No. PV6274
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human CDC7 and DBF4 proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for the subunits (CD7/DBF4) are NP_003494.1 and NP_006707.1.

CDC7, or Cell Division Cycle 7, is a serine/threonine kinase that plays a crucial role in the initiation of DNA replication. It is essential for the activation of replication origins during the S phase of the cell cycle. ASK, also known as Activator of S Phase Kinase/DBF4, is a regulatory subunit that binds to CDC7 and activates its kinase activity. The binding of ASK to CDC7 is necessary for the proper functioning of CDC7, as it regulates the activation and specificity of the kinase. Together, the CDC7/ASK complex is critical for the regulation of DNA replication. This complex phosphorylates components of the pre-replication complex at replication origins, thereby promoting the initiation of DNA synthesis. By ensuring that replication origins fire at the appropriate time, the CDC7/ASK complex helps to maintain genomic stability and proper cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00311, Q9UBU7
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.25mM DTT, 25% glycerol, 0.1mM EDTA, 10mM glutathione, 0.1mM PMSF, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 10926, 8317
Molecular Weight (g/mol) 91.2-103.5 kDa
Name Human CDC7/DBF4, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI597260; Cdc7; CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1; Cdc7 Kinase; Cdc7l1; CDC7-like 1; CDC7-related kinase; cell division cycle 7; cell division cycle 7 (S. cerevisiae); cell division cycle 7 homolog; cell division cycle 7-like 1; cell division cycle 7-like protein 1; cell division cycle 7-related protein kinase; HsCDC7; Hsk1; huCDC7; MGC117361; MGC126237; MGC126238; MGC91067; muCdc7
Common Name CDC7/ASK
Gene Symbol CDC7, DBF4
Protein Length 1-574
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-574
Content And Storage Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Assay Entry Biochemical binding/kinetic
Kinase Group Serine/Threonine Kinase
Target Entry CDC7/DBF4
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Cell Cycle & Proliferation, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.