Learn More
Medchemexpress LLC HY-101523 10mg Medchemexpress, Cdc7-IN-1 CAS:1402055-25-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10152310MG
Encompass_Preferred
Medchemexpress, HY-101523 10mg Cdc7-IN-1 CAS:1402055-25-7 Cdc7-IN-1 (Compound 13) is a highly potent, selective and ATP competitive inhibitor of Cdc7 kinase, with an IC50 value of 0.6 nM at 1 mM ATP and with slow off-rate characteristics. Cdc7-IN-1 potently inhibits Cdc7 activity in cancer cells, and effectively induces cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7855