Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdc7-IN-5 | 1402057-86-6 | 96.0% | 100 MG
SDP

Catalog No. 5000338610
Encompass_Preferred

Cdc7-IN-5 is a potent Cdc7 kinase inhibitor. This compound is extracted from patent WO2019165473A1 and targets Cdc7, a serine-threonine protein kinase enzyme essential for initiating DNA replication in the cell cycle.

  • Potent Cdc7 kinase inhibitor
  • Extracted from patent WO2019165473A1
  • Targets serine-threonine protein kinase enzyme Cdc7
  • Essential for DNA replication initiation
  • Appearance: solid, white to light yellow
  • Molecular weight: 445.47
  • Molecular formula: C25H23N3O5

Catalog No. 50-003-38610 Supplier Medchemexpress LLC Supplier No. HY130517100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cdc7-IN-5 is a potent Cdc7 kinase inhibitor. This compound is extracted from patent WO2019165473A1 and targets Cdc7, a serine-threonine protein kinase enzyme essential for initiating DNA replication in the cell cycle.

  • Potent Cdc7 kinase inhibitor
  • Extracted from patent WO2019165473A1
  • Targets serine-threonine protein kinase enzyme Cdc7
  • Essential for DNA replication initiation
  • Appearance: solid, white to light yellow
  • Molecular weight: 445.47
  • Molecular formula: C25H23N3O5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.