Learn More
Medchemexpress LLC Cdc7-IN-5 (compound I-B) | 1402057-86-6 | 96.7% | C25H23N3O5 | 10MG
SDP
Catalog No. 5000260882
Encompass_Preferred
Cdc7-IN-5 is a small-molecule inhibitor of the CDC7 serine-threonine kinase used in research on DNA replication and cell cycle regulation. The compound is provided as a dry solid with molecular formula C25H23N3O5, molecular weight 445.47, and reported purity around 96.7%.
  • Inhibits CDC7 kinase activity in biochemical and cellular assays.
  • Useful for studies of DNA replication and cell cycle control.
  • Supplied as a dry solid with high reported purity for research use.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in research-scale quantities, including 10 mg and sample sizes.
Catalog No. 50-002-60882 Supplier Medchemexpress LLC Supplier No. HY13051710MG
Cdc7-IN-5 is a small-molecule inhibitor of the CDC7 serine-threonine kinase used in research on DNA replication and cell cycle regulation. The compound is provided as a dry solid with molecular formula C25H23N3O5, molecular weight 445.47, and reported purity around 96.7%.
  • Inhibits CDC7 kinase activity in biochemical and cellular assays.
  • Useful for studies of DNA replication and cell cycle control.
  • Supplied as a dry solid with high reported purity for research use.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Available in research-scale quantities, including 10 mg and sample sizes.