Learn More
TARGETMOL CHEMICALS INC CDC7-IN-7C 5MG
SDP
Catalog No. 502580842
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cdc7-IN-7c (Cdc7 inhibitor-7c) has antitumor activity and has inhibitory effect on liver cancer lung cancer kidney cancer brain cancer and cervical cancer. purity: 99%
Catalog No. 50-258-0842 Supplier TARGETMOL CHEMICALS INC Supplier No. T238675MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cdc7-IN-7c (Cdc7 inhibitor-7c) has antitumor activity and has inhibitory effect on liver cancer lung cancer kidney cancer brain cancer and cervical cancer. purity: 99%