Learn More
Aviva Systems Biology Corp CDC73 Antibody - C-terminal region (ARP63639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDC73. It was validated on Western Blot. . Alternate names: C1orf28, FLJ23316, HPTJT, HRPT2, HYXApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-039