Learn More
Hach Company Radiometer Analytical CDC745 Conductivity Cell (2-pole Pt sensor, k = 1.0 cm-1, epoxy body, screw cap)
Catalog No. 501997614 Shop All Hach Company Products
Encompass
The Radiometer CDC745 is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1) and robust epoxy body. The CDC745 sensor's 2 platinum plate design is ideal for routine conductivity measurements in various aqueous and non-aqueous media. The CDC745 has a screw cap connection and requires the CL136 cable (part no. A94L136) for compatibility with Radiometer Analytical CDM210/230/92 conductivity meters or the CL143 cable (part no. A94L143) for compatibilty with Hach sensION+ benchtop conductivity meters.
Catalog No. 50-199-7614 Supplier Hach Company Supplier No. E61M013
The Radiometer CDC745 is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1) and robust epoxy body. The CDC745 sensor's 2 platinum plate design is ideal for routine conductivity measurements in various aqueous and non-aqueous media. The CDC745 has a screw cap connection and requires the CL136 cable (part no. A94L136) for compatibility with Radiometer Analytical CDM210/230/92 conductivity meters or the CL143 cable (part no. A94L143) for compatibilty with Hach sensION+ benchtop conductivity meters.