Learn More
Aviva Systems Biology Corp CDCA7L Antibody - middle region (ARP57292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDCA7L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDCA7L. Alternate names: DKFZp762L0311, JPO2, R1, RAM2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53994