Learn More
Aat Bioquest CDCF [5-(and-6)-Carboxy-2',7'-dichlorofluorescein] *Mixed isomers*
SDP
Supplier:  Aat Bioquest 235
Encompass_Preferred
CDCF [5(6)-carboxy-2',7'-dichlorofluorescein] is the fluorescent product of 5(6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate and 5(6)-carboxy-2',7'-dichlorofluorescein diacetate in cells.
Catalog No. 50-195-2815