Learn More
Medchemexpress LLC CDCP1 Mouse Biotin 50ug | 50ug
SDP
Catalog No. 5000187620
Encompass_Preferred
The CDCP1 protein has been implicated in cell adhesion and matrix association suggesting a role in important cellular interactions Its phosphorylation suggests a dynamic function in regulating anchorage migration proliferation or differentiation CDCP1 Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived CDCP1 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87620 Supplier Medchemexpress LLC Supplier No. HYP7810150UG
The CDCP1 protein has been implicated in cell adhesion and matrix association suggesting a role in important cellular interactions Its phosphorylation suggests a dynamic function in regulating anchorage migration proliferation or differentiation CDCP1 Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived CDCP1 protein expressed by HEK293 with C-Avi C-His labeled tag