Learn More
Medchemexpress LLC HY-100901 5mg Medchemexpress, CDD3505 CAS:173865-33-3 Purity:>98%
SDP
Catalog No. 501931992
Encompass_Preferred
Medchemexpress, HY-100901 5mg CDD3505 CAS:173865-33-3 CDD3505 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1992 Supplier Medchemexpress LLC Supplier No. HY1009015MG
Medchemexpress, HY-100901 5mg CDD3505 CAS:173865-33-3 CDD3505 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.