Learn More
Medchemexpress LLC HY-100902 5mg Medchemexpress, CDD3506 CAS:197913-15-8 Purity:>98%
SDP
Catalog No. 501931993
Encompass_Preferred
Medchemexpress, HY-100902 5mg CDD3506 CAS:197913-15-8 CDD3506 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1993 Supplier Medchemexpress LLC Supplier No. HY1009025MG
Medchemexpress, HY-100902 5mg CDD3506 CAS:197913-15-8 CDD3506 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.